AMBH580643FD Ambeed, Inc.

Methyl 5-chloro-4-fluorothiophene-2-carboxylate

  • MDL number MFCD30189030
  • Cas Number : 2089319-62-8
  • Molecular Weight : 194.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/2089319-62-8.html

Safety Information

MSDS https://www.ambeed.com/products/2089319-62-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart