AMBH58064408 Ambeed, Inc.

5-(3-(4-(4-Chlorophenyl)piperidin-1-yl)propyl)pyrrolo[1,2-a]quinoxalin-4(5H)-one

  • MDL number MFCD30489012
  • Cas Number : 1380392-05-1
  • Molecular Weight : 419.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1380392-05-1.html

Safety Information

MSDS https://www.ambeed.com/products/1380392-05-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart