AMBH5806440D Ambeed, Inc.

(4,5-Dichloro-1,2-phenylene)dimethanol

  • MDL number MFCD06656516
  • Cas Number : 24006-92-6
  • Molecular Weight : 207.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/24006-92-6.html

Safety Information

MSDS https://www.ambeed.com/products/24006-92-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart