AMBH58064410 Ambeed, Inc.

2-Bromonaphthalen-1-amine

  • MDL number MFCD11870111
  • Cas Number : 771-14-2
  • Molecular Weight : 222.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/771-14-2.html

Safety Information

MSDS https://www.ambeed.com/products/771-14-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart