AMBH58064414 Ambeed, Inc.

6-Chloro-3,3-dimethyl-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine

  • MDL number MFCD28399436
  • Cas Number : 1403899-44-4
  • Molecular Weight : 182.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1403899-44-4.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1403899-44-4.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart