AMBH58064417 Ambeed, Inc.

2-(Dimethylamino)-2-phenylacetic acid

  • MDL number MFCD00957245
  • Cas Number : 14758-99-7
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/14758-99-7.html

Safety Information

MSDS https://www.ambeed.com/products/14758-99-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart