AMBH58064418 Ambeed, Inc.

tert-Butyl (4-bromobutan-2-yl)carbamate

  • MDL number MFCD22576327
  • Cas Number : 128412-15-7
  • Molecular Weight : 252.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (4-bromobutan-2-yl)carbamate
Inchi Code 1S/C9H18BrNO2/c1-7(5-6-10)11-8(12)13-9(2,3)4/h7H,5-6H2,1-4H3,(H,11,12)
Inchi Key BLROSIRAEKEFNI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/128412-15-7.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/128412-15-7.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart