AMBH5806441A Ambeed, Inc.

8-Aminoquinolin-6-ol

  • MDL number MFCD06408351
  • Cas Number : 7402-16-6
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/7402-16-6.html

Safety Information

signalword Warning
pcodes P233-P260-P261-P264-P271-P280-P302+P352-P304-P304+P340-P305+P351+P338-P312-P321-P332+P313-P337+P313-P340-P362-P403-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/7402-16-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart