AMBH58064424 Ambeed, Inc.

4-Hydroxyisophthalaldehyde

  • MDL number MFCD00003334
  • Cas Number : 3328-70-9
  • Molecular Weight : 150.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109°C at N/A mmHg
Purity 98
Flash Point 128.9
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 266°C at 760 mmHg
Product Link https://www.ambeed.com/products/3328-70-9.html

Safety Information

signalword Warning
pcodes P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313
MSDS https://www.ambeed.com/products/3328-70-9.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart