AMBH58064427 Ambeed, Inc.

2,2-Bis[(4S)-4-benzyl-2-oxazolin-2-yl]propane

  • MDL number MFCD30343613
  • Cas Number : 176706-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/176706-98-2.html

Safety Information

MSDS https://www.ambeed.com/products/176706-98-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart