AMBH58064432 Ambeed, Inc.

Benzofuroxan

  • MDL number MFCD00005807
  • Cas Number : 480-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/480-96-6.html

Safety Information

MSDS https://www.ambeed.com/products/480-96-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart