AMBH58064433 Ambeed, Inc.

3-(1H-Imidazol-4-yl)acrylic acid

  • MDL number MFCD00005203
  • Cas Number : 104-98-3
  • Molecular Weight : 138.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/104-98-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/104-98-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart