AMBH5806444B Ambeed, Inc.

2,5-Bis(trifluoromethyl)-1,3,4-oxadiazole

  • MDL number N/A
  • Cas Number : 1868-48-0
  • Molecular Weight : 206.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20°C
Physical Form liquid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 54.3°C at N/A mmHg
Product Link https://www.ambeed.com/products/1868-48-0.html

Safety Information

signalword Warning
pcodes P210-P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1868-48-0.html
pictograms GHS07
hstatements H227-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart