AMBH5806444D Ambeed, Inc.

tert-Butyl 4-(3-(dimethylamino)acryloyl)piperidine-1-carboxylate

  • MDL number MFCD08898850
  • Cas Number : 960201-86-9
  • Molecular Weight : 282.3785

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form Solid or liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/960201-86-9.html

Safety Information

signalword Danger
pcodes P261-P280-P273-P333+P313-P391
MSDS https://www.ambeed.com/products/960201-86-9.html
pictograms GHS07,GHS09
hstatements H302-H315-H319-H335-H410
Availability details for
Quantity
  • Availability
  • Add to Cart