AMBH58064459 Ambeed, Inc.

Gallium selenide

  • MDL number MFCD00016103
  • Cas Number : 12024-24-7
  • Molecular Weight : 376.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 937�t N/A mmHg
Iupac Name gallium(III) selenide
Inchi Code 1S/2Ga.3Se/q2*+3;3*-2
Inchi Key ZZEMEJKDTZOXOI-UHFFFAOYSA-N

Description

Melting Point 937�t N/A mmHg
Purity 99%
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/12024-24-7.html

Safety Information

signalword Danger
pcodes P260-P273-P284-P310
MSDS https://www.ambeed.com/products/12024-24-7.html
pictograms GHS06,GHS09
hstatements H301-H330-H400
Availability details for
Quantity
  • Availability
  • Add to Cart