AMBH5806445F Ambeed, Inc.

N-Benzyl-N-ethylethanamine

  • MDL number MFCD00026731
  • Cas Number : 772-54-3
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 208.3�t 760 mmHg
Iupac Name N-benzyl-N-ethylethanamine
Inchi Code 1S/C11H17N/c1-3-12(4-2)10-11-8-6-5-7-9-11/h5-9H,3-4,10H2,1-2H3
Inchi Key ZWRDBWDXRLPESY-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 208.3�t 760 mmHg
Product Link https://www.ambeed.com/products/772-54-3.html

Safety Information

signalword Danger
pcodes P210-P260-P264-P280-P301+P330+P331+P310-P303+P361+P353+P310+P363-P304+P340+P310-P305+P351+P338+P310-P370+P378-P403+P235-P405-P501
MSDS https://www.ambeed.com/products/772-54-3.html
pictograms GHS05
hstatements H227-H314
Availability details for
Quantity
  • Availability
  • Add to Cart