AMBH58064463 Ambeed, Inc.

T863

  • MDL number MFCD11977270
  • Cas Number : 701232-20-4
  • Molecular Weight : 394.4669

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/701232-20-4.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/701232-20-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart