AMBH5806446C Ambeed, Inc.

Potassium O-pentyl carbonodithioate

  • MDL number MFCD00059150
  • Cas Number : 2720-73-2
  • Molecular Weight : 202.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 21°C at N/A mmHg
Purity 97
Flash Point 137
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/2720-73-2.html

Safety Information

signalword Danger
pcodes P210-P240-P241-P261-P264-P280-P332+P313-P302+P352-P305+P351+P338-P312
hcodes 4.1
MSDS https://www.ambeed.com/products/2720-73-2.html
pictograms GHS02,GHS07
hstatements H228-H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart