AMBH5806446D Ambeed, Inc.

4-Bromo-2-nitro-6-(trifluoromethoxy)aniline

  • MDL number MFCD16628052
  • Cas Number : 1257535-31-1
  • Molecular Weight : 301.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1257535-31-1.html

Safety Information

MSDS https://www.ambeed.com/products/1257535-31-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart