AMBH58064471 Ambeed, Inc.

(2R,5S)-2,5-Diaminohexanedioic acid dihydrochloride

  • MDL number MFCD09836085
  • Cas Number : 213686-09-0
  • Molecular Weight : 249.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/213686-09-0.html

Safety Information

MSDS https://www.ambeed.com/products/213686-09-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart