AMBH58064476 Ambeed, Inc.

5-Fluoro-2-oxo-1,2-dihydropyridine-3-carboxamide

  • MDL number MFCD29922097
  • Cas Number : 1956340-92-3
  • Molecular Weight : 156.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1956340-92-3.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/1956340-92-3.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart