AMBH58064477 Ambeed, Inc.

4-(Aminomethyl)-3-fluorobenzonitrile

  • MDL number MFCD09943431
  • Cas Number : 701264-00-8
  • Molecular Weight : 150.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/701264-00-8.html

Safety Information

MSDS https://www.ambeed.com/products/701264-00-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart