AMBH5806447D Ambeed, Inc.

6-Chloro-2-(chloromethyl)-1H-benzo[d]imidazole

  • MDL number MFCD00060503
  • Cas Number : 20443-38-3
  • Molecular Weight : 201.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 404.6�t 760 mmHg
Iupac Name 6-chloro-2-(chloromethyl)-1H-benzimidazole
Inchi Code 1S/C8H6Cl2N2/c9-4-8-11-6-2-1-5(10)3-7(6)12-8/h1-3H,4H2,(H,11,12)
Inchi Key CJNALUZDTPETCS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Cold-chain transportation
Country of Origin CN
Boiling point 404.6�t 760 mmHg
Product Link https://www.ambeed.com/products/20443-38-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/20443-38-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart