AMBH58064488 Ambeed, Inc.

6-Chloro-N-methylpicolinamide

  • MDL number MFCD09862945
  • Cas Number : 845306-04-9
  • Molecular Weight : 170.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8°C
Physical Form Light-yellow to yellow solid or liquid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/845306-04-9.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/845306-04-9.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart