AMBH5806448E Ambeed, Inc.

Benzyl 4-methyl-5-oxo-1,4-diazepane-1-carboxylate

  • MDL number MFCD28347727
  • Cas Number : 1359072-90-4
  • Molecular Weight : 262.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1359072-90-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1359072-90-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart