AMBH5806448F Ambeed, Inc.

4-Bromo-2-methyl-6-nitrophenol

  • MDL number MFCD09835004
  • Cas Number : 20294-50-2
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 278.5�t 760 mmHg
Iupac Name 4-bromo-2-methyl-6-nitrophenol
Inchi Code 1S/C7H6BrNO3/c1-4-2-5(8)3-6(7(4)10)9(11)12/h2-3,10H,1H3
Inchi Key KWNYDIOUTKEIFL-UHFFFAOYSA-N

Description

Synonym N/A
Purity 95%
Flash Point 122.2
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 278.5�t 760 mmHg
Product Link https://www.ambeed.com/products/20294-50-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/20294-50-2.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart