AMBH58064490 Ambeed, Inc.

Canagliflozin

  • MDL number MFCD18251436
  • Cas Number : 842133-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/842133-18-0.html

Safety Information

MSDS https://www.ambeed.com/products/842133-18-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart