AMBH58064492 Ambeed, Inc.

5-Bromo-3-(trifluoromethyl)picolinaldehyde

  • MDL number MFCD16606468
  • Cas Number : 1227489-83-9
  • Molecular Weight : 254.0040

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form Liquid or Solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1227489-83-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1227489-83-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart