AMBH58064493 Ambeed, Inc.

Asciminib

  • MDL number MFCD31560488
  • Cas Number : 1492952-76-7
  • Molecular Weight : 449.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1492952-76-7.html

Safety Information

MSDS https://www.ambeed.com/products/1492952-76-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart