AMBH58064495 Ambeed, Inc.

NIBR189

  • MDL number MFCD13304236
  • Cas Number : 1599432-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1599432-08-2.html

Safety Information

MSDS https://www.ambeed.com/products/1599432-08-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart