AMBH58064499 Ambeed, Inc.

5,8-Dibromoquinoline

  • MDL number MFCD04966994
  • Cas Number : 81278-86-6
  • Molecular Weight : 286.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,8-dibromoquinoline
Inchi Code 1S/C9H5Br2N/c10-7-3-4-8(11)9-6(7)2-1-5-12-9/h1-5H
Inchi Key LCSHATKNSDVDRY-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/81278-86-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/81278-86-6.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart