AMBH5806449B Ambeed, Inc.

3-Ethynylquinoline

  • MDL number MFCD03286159
  • Cas Number : 78593-40-5
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/78593-40-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/78593-40-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart