AMBH580644A5 Ambeed, Inc.

(R)-tert-Butyl (1-hydroxypent-4-yn-2-yl)carbamate

  • MDL number MFCD09800406
  • Cas Number : 1217637-01-8
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1217637-01-8.html

Safety Information

MSDS https://www.ambeed.com/products/1217637-01-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart