AMBH580644A9 Ambeed, Inc.

Magnesium oxide

  • MDL number MFCD00011109
  • Cas Number : 1309-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/1309-48-4.html

Safety Information

MSDS https://www.ambeed.com/products/1309-48-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart