AMBH580644AB Ambeed, Inc.

Methyl 6-bromo-3-chloropicolinate

  • MDL number MFCD14698108
  • Cas Number : 350602-14-1
  • Molecular Weight : 250.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/350602-14-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/350602-14-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart