AMBH580644AF Ambeed, Inc.

Valbenazine

  • MDL number MFCD28963976
  • Cas Number : 1025504-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1025504-45-3.html

Safety Information

MSDS https://www.ambeed.com/products/1025504-45-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart