AMBH580644B2 Ambeed, Inc.

N1,N1-Bis(3-(Dimethylamino)propyl)-N3,N3-dimethylpropane-1,3-diamine

  • MDL number MFCD00190091
  • Cas Number : 33329-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/33329-35-0.html

Safety Information

MSDS https://www.ambeed.com/products/33329-35-0.html
Availability details for
Quantity
  • Availability
  • Add to Cart