AMBH580644B3 Ambeed, Inc.

2-(3-Methoxy-2-methylphenyl)acetic acid

  • MDL number MFCD18398342
  • Cas Number : 33797-36-3
  • Molecular Weight : 180.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/33797-36-3.html

Safety Information

MSDS https://www.ambeed.com/products/33797-36-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart