AMBH580644BB Ambeed, Inc.

1,4-Dipropylbenzene

  • MDL number MFCD03844789
  • Cas Number : 4815-57-0
  • Molecular Weight : 162.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 84.3
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/4815-57-0.html

Safety Information

signalword Warning
pcodes P210-P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/4815-57-0.html
pictograms GHS07
hstatements H227-H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart