AMBH580644BE Ambeed, Inc.

tert-Butyl 3-hydroxy-4-oxopiperidine-1-carboxylate

  • MDL number MFCD18157599
  • Cas Number : 1130156-23-8
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 160.6
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 341.9�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1130156-23-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1130156-23-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart