AMBH580644C1 Ambeed, Inc.

Isoborneol

  • MDL number MFCD00074821
  • Cas Number : 124-76-5
  • Molecular Weight : 154.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 208-214�t N/A mmHg
bp 212�t 760 mmHg
Iupac Name 1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Inchi Code 1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3
Inchi Key DTGKSKDOIYIVQL-UHFFFAOYSA-N

Description

Melting Point 208-214�t N/A mmHg
Purity 90%
Flash Point 65.6
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 212�t 760 mmHg
Product Link https://www.ambeed.com/products/124-76-5.html

Safety Information

signalword Danger
pcodes P210
MSDS https://www.ambeed.com/products/124-76-5.html
pictograms GHS02,GHS07
hstatements H228-H315
Availability details for
Quantity
  • Availability
  • Add to Cart