AMBH580644C5 Ambeed, Inc.

(R)-2-(7-(4-Fluoro-N-methylphenylsulfonamido)-6,7,8,9-tetrahydropyrido[1,2-a]indol-10-yl)acetic acid

  • MDL number MFCD22741619
  • Cas Number : 1218918-62-7
  • Molecular Weight : 416.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1218918-62-7.html

Safety Information

MSDS https://www.ambeed.com/products/1218918-62-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart