AMBH580644CE Ambeed, Inc.

3-((Dimethylamino)methyl)-5-methylhexan-2-one

  • MDL number MFCD22683881
  • Cas Number : 91342-74-4
  • Molecular Weight : 171.2798

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/91342-74-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/91342-74-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart