AMBH580644D1 Ambeed, Inc.

4,6-Dimethoxypyridin-3-amine

  • MDL number MFCD09027594
  • Cas Number : 89943-34-0
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/89943-34-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/89943-34-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart