AMBH580644D4 Ambeed, Inc.

Ethyl 2-bromo-5-methylthiazole-4-carboxylate

  • MDL number MFCD01924782
  • Cas Number : 56355-62-5
  • Molecular Weight : 250.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/56355-62-5.html

Safety Information

MSDS https://www.ambeed.com/products/56355-62-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart