AMBH580644DC Ambeed, Inc.

1-(Amino(phenyl)methyl)cyclopropanol

  • MDL number MFCD22681561
  • Cas Number : 1391737-71-5
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1391737-71-5.html

Safety Information

MSDS https://www.ambeed.com/products/1391737-71-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart