AMBH580644DF Ambeed, Inc.

2,2-Thiodiethylene Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]

  • MDL number MFCD00059346
  • Cas Number : 41484-35-9
  • Molecular Weight : 642.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/41484-35-9.html

Safety Information

signalword Danger
pcodes P273
MSDS https://www.ambeed.com/products/41484-35-9.html
pictograms GHS09
hstatements H400-H411
Availability details for
Quantity
  • Availability
  • Add to Cart