AMBH580644E8 Ambeed, Inc.

Strontium oxalate

  • MDL number MFCD00050803
  • Cas Number : 814-95-9
  • Molecular Weight : 175.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/814-95-9.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/814-95-9.html
pictograms GHS07
hstatements H302-H312
Availability details for
Quantity
  • Availability
  • Add to Cart