AMBH580644EA Ambeed, Inc.

3-Amino-6-methoxypicolinic acid

  • MDL number MFCD09264292
  • Cas Number : 870971-19-0
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 384.7�t 760 mmHg
Iupac Name 3-amino-6-methoxy-2-pyridinecarboxylic acid
Inchi Code 1S/C7H8N2O3/c1-12-5-3-2-4(8)6(9-5)7(10)11/h2-3H,8H2,1H3,(H,10,11)
Inchi Key DDAPSAVXCBGGBC-UHFFFAOYSA-N

Description

Synonym N/A
Purity 97%
Flash Point 214.7
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 384.7�t 760 mmHg
Product Link https://www.ambeed.com/products/870971-19-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/870971-19-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart