AMBH580644F1 Ambeed, Inc.

(2'-Fluoro-[1,1'-biphenyl]-4-yl)boronic acid

  • MDL number MFCD03095260
  • Cas Number : 1383531-49-4
  • Molecular Weight : 216.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point N/A
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1383531-49-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1383531-49-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart